BRZMBRCFLEX4
Mobius® Breez 2 mL Microbioreactor
CFlex Tubing, Qty 4
Synonym(s):
Mobius® Breez 2 mL Microbioreactor with CFlex Tubing, Qty 4
Select a Size
+61 2 98410588or
This item is not able to be purchased as a guest user. For further information, please contact us at .Select a Size
About This Item
+61 2 98410588or
This item is not able to be purchased as a guest user. For further information, please contact us at .Recommended Products
sterility
γ-irradiated
technique(s)
cell culture | mammalian: suitable
volume
2 mL
Looking for similar products? Visit Product Comparison Guide
Application
- Monoclonal antibodies
- Recombinant proteins
Features and Benefits
For perfusion cell line development, media screening and optimization, and early process development, the ability to run multiple experiments at rapid speed is essential. The Mobius® Breez Microbioreactor System is designed for rapid cell culture experiments, providing an easy-to use solution to produce high quality datasets. It allows for increased productivity and shorter development timelines.
Other Notes
The pharmaceutical and biotechnology industries are highly regulated. To support you as you navigate this challenging environment, we offer a wide range of services that can help save time and lower costs. All of our services are performed by global experts who have an intimate knowledge of our equipment backed by decades of experience, giving you peace of mind at every step.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page summarizes several performance attributes and scalability data for Mobius® iFlex Bioreactors, single-use, stirred tank systems designed to support traditional and intensified fed-batch and perfusion cell culture applications.
Related Content
The Mobius® Breez Microbioreactor is a 2 mL automated single-use perfusion system designed to support gentle, adaptable, and reproducible cell processes.
Mobius® iFlex Bioreactor family offers single-use bioreactors for mammalian cell culturing from development to production.
This page describes the use of a small-scale perfusion bioreactor with integrated microfluidic flow control and sensor technologies to predict large-scale processes.
Learn more on mAB bioreactor production, and the products you need for this step.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service