Skip to Content
Merck
All Photos(1)

Key Documents

BM-1SP

Millipore

Auto2D® Solution Chip Plus

Auto2D® 2-D Electrophoresis Device

Synonym(s):

2D Electrophoresis, 2DE, Electrode, IEF, SDS-PAGE, Solution Chip, Solution Chip Plus, Tray

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$190.00

$190.00


Estimated to ship on23 May 2025



Select a Size

Change View
1 PKG
$190.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

$190.00


Estimated to ship on23 May 2025


product line

Auto2D®

compatibility

Auto2D® 2-D Electrophoresis Device

storage temp.

room temp

General description

Solution Chip Plus is used for applying solution on the Auto2D® 2-D Electrophoresis Device when running the system with "Auto2D Plus" mode.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service