ATBHPRE01
M Air T Pressure Reducer
The M Air T Pressure Reducer helps to regulate the gas pressure in a line to be monitored.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104008
eCl@ss:
25140604
NACRES:
NB.11
manufacturer/tradename
M Air T™
input
sample type pharmaceutical(s)
application(s)
environmental
compatibility
for use with M Air T™ Compressed Gas and Air Test Kit
shipped in
ambient
Legal Information
M AIR T is a trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service