Skip to Content
Merck
All Photos(1)

Key Documents

AQ503F

Sigma-Aldrich

Goat Anti-Human IgM+IgG+IgA Antibody, F(ab′)2, FITC conjugate

0.5 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$230.00

$230.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$230.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$230.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

0.5 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is the most abundant immunoglobulin comprising two heavy chains and two light chains. Each molecule has two antigen binding sites. IgG has 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%). IgM constitutes about 10% of serum immunoglobulins. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. Monomeric IgA constitutes 5-15 % of the serum immunoglobulins whereas dimeric IgA is localized to mucosa surfaces.

Specificity

Reacts with the heavy and light chains of human IgM, IgG and IgA, and with the light chains of human IgD.
The antibody reacts with the heavy and light chains of human IgM, IgG and IgA, and with the light chains of human IgD as demonstrated by ELISA and flow cytometry.

Immunogen

Prepared from pepsin digest of human Ig.

Application

Goat anti-Human IgM+IgG+IgA Antibody, F(ab′)2, FITC conjugate detects level of Human IgM+IgG+IgA & has been published & validated for use in ELISA & IF.
Immunofluorescence: ≤1 μg/106 cells.
ELISA

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

Gel filtration chromatography
Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0 mL PBS/NaN3.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Obuchowicz et al.
Frontiers in pharmacology, 10, 1687-1687 (2020-02-23)
Here, we examine the effects of prenatal administration of two antidepressants-imipramine (IMI) and venlafaxine (VEN)-on morphology and activity of a primary glial culture. Microglia are targeted by antidepressants used for antenatal depression and are important regulators of central nervous system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service