Skip to Content
Merck
All Photos(1)

Key Documents

AQ502

Sigma-Aldrich

Goat Anti-Mouse IgM+IgG+IgA Antibody, F(ab′)2, Species Adsorbed

0.5 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$174.00

$174.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$174.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$174.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

0.5 mg/mL

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is the most abundant immunoglobulin comprising two heavy chains and two light chains. Each molecule has two antigen binding sites. IgG has 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%). IgM constitutes about 10% of serum immunoglobulins. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. Monomeric IgA constitutes 5-15 % of the serum immunoglobulins whereas dimeric IgA is localized to mucosa surfaces.

Specificity

Reacts with heavy and light chains of mouse IgM, IgG1, IgG2a, IgG2b, IgG3 and IgA. Absorbed for human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.
The antibody reacts with the heavy and light chains of mouse IgM, IgG1, IgG2a, IgG2b, IgG3 and IgA as demonstrated by ELISA and flow cytometry. Minimal cross reactivity with human immunoglobulins.

Immunogen

Epitope: Mouse IgM, IgG, IgA
Prepared from pepsin digest of mouse Ig (H+L).

Application

Goat anti-Mouse IgM+IgG+IgA Antibody, F(ab′)2, Species Adsorbed is an antibody against Mouse IgM+IgG+IgA for use in ELISA.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

Gel filtration chromatography
Purified by gel filtration chromatography of pepsin digested antibody. Liquid in 1.0 mL of 100 mM borate buffered saline, pH 8.2.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service