Skip to Content
Merck
All Photos(1)

Key Documents

AQ160

Sigma-Aldrich

Rabbit Anti-Mouse IgG Antibody, F(ab′)2

1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$241.00

$241.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$241.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$241.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with mouse IgG gamma chain, as well as the light chains from all mouse immunoglobulin classes. Monospecific by IEP against whole mouse serum.

Immunogen

Highly purified mouse IgG isolated from pooled normal mouse serum.

Application

Optimal working dilutions must be determined by end user.
Rabbit anti-Mouse IgG Antibody, F(ab′)2 is an antibody against Mouse IgG for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Quality

Pepsin digest shows a single band on IEP at an antigen concentration of 10 mg/mL when run against anti-mouse IgG (H&L) and anti-F(ab′)2. No reaction was observed against anti-rabbit IgG (Fc).

Physical form

Affinity purified immunoglobulin, F(ab′)2 fragment. Liquid in 0.02 M Phosphate Buffer, 0.25 M NaCl, 0.1% sodium azide.

Storage and Stability

Store at +2-8°C for up to six months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean C Rousseau et al.
BMC musculoskeletal disorders, 9, 74-74 (2008-05-30)
Rheumatoid arthritis (RA) is a chronic auto-immune disease with extensive articular cartilage destruction. Aggrecan depletion, mediated by aggrecanases is one of the first signs of early cartilage erosion. We investigated, whether measurement of aggrecan and fragments thereof in serum, could

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service