Skip to Content
Merck
All Photos(1)

Key Documents

AQ132B

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, F(ab′)2, biotin-SP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$263.00

$263.00


Estimated to ship on02 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
$263.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$263.00


Estimated to ship on02 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, F(ab′)2, biotin-SP conjugate validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoît Silvestre de Ferron et al.
Addiction biology, 22(6), 1870-1882 (2016-10-26)
Low to moderate perinatal ethanol exposure (PEE) may have disastrous consequences for the central nervous system resulting notably in permanent cognitive deficits. Learning and memory are mediated in the hippocampus by long-term potentiation (LTP) and long term depression (LTD), two
Raquel Martínez-Vega et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(2), 418-432 (2014-11-12)
Nutritional imbalance is emerging as a causative factor of hearing loss. Epidemiologic studies have linked hearing loss to elevated plasma total homocysteine (tHcy) and folate deficiency, and have shown that folate supplementation lowers tHcy levels potentially ameliorating age-related hearing loss.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service