Skip to Content
Merck
All Photos(1)

Key Documents

AQ112P

Sigma-Aldrich

Goat Anti-Human IgG Antibody, F(ab′)2, HRP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$347.00

$347.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
$347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$347.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Goat anti-Human IgG Antibody, F(ab′)2, HRP conjugate detects level of Human IgG & has been published & validated for use in ELISA & WB.

Storage and Stability

Reconstitution: Store freeze-dried product at 4°C until opened for up to 12 months. Reconstitute to 1 mg/mL with distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. (To judge clarity, draw product into a pasteur pipette). Product is stable for several weeks at 4°C as an undiluted liquid. Discard dilutions after more than one day. For extended storage after reconsitutiton, add an equal volume of glycerol to make a final concentation of 50% glycerol and store at -20°C. Please remember that the concentration of protein (and buffer salts) will decrease to one-half of the original after addition of glycerol.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmelo Carmona-Rivera et al.
JCI insight, 5(13) (2020-06-03)
Rheumatoid arthritis (RA) is characterized by synovial joint inflammation, cartilage damage, and dysregulation of the adaptive immune system. While neutrophil extracellular traps (NETs) have been proposed to play a role in the generation of modified autoantigens and in the activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service