Skip to Content
Merck
All Photos(1)

Key Documents

AP506A

Sigma-Aldrich

Goat Anti-Human Ig λ chain Antibody, Alkaline Phosphatase conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$307.00

$307.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$307.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$307.00


Estimated to ship on05 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and lambda light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Reacts with human lambda light chains. Absorbed for human myeloma proteins with kappa light chains.
The antibody reacts with human lambda light chains as demonstrated by ELISA and flow cytometry.

Immunogen

Epitope: Ig lambda chain
Prepared from pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains.

Application

ELISA: 1:2,000-1:4,000.

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
This Goat anti-Human Ig λ chain Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA for the detection of Human Ig λ chain.

Physical form

Affinity Chromatography
Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. For long term storage, aliquote and store at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service