Skip to Content
Merck
All Photos(1)

Key Documents

AP200B

Sigma-Aldrich

Goat Anti-Mouse light chain Antibody, biotin conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$318.00

$318.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$318.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$318.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and gamma light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, horse, human, rabbit, rat, and sheep.
The antibody reacts strongly with native primary antibodies primarily with kappa light chains. It is not suitable for detecting lambda light chains. The antibody does not react with the heavy chain of mouse IgG. The antibody has been tested by ELISA and adsorbed to ensure minimal cross-reaction with bovine, goat, horse, human, rabbit, rat, and sheep immunoglobulins.

Immunogen

Epitope: Kappa light chain
Prepared from purified mouse IgG light chain.

Application

Suggested dilutions:
Western Blotting: 1:20,000 - 1:400,000 of the 1 mg/mL stock
ELISA: 1:20,000 - 1:400,000 of the 1 mg/mL stock
Immunohistochemistry: 1:500 - 1:5,000 of the 1 mg/mL stock
Immunocytochemistry: 1:500 - 1:5,000 of the 1 mg/mL stock
Flow Cytometry: 1:200 - 1:1,000 of the 1 mg/mL stock

Optimal working dilutions must be determined by the end user.
Detect Mouse light chain using this Goat anti-Mouse light chain Antibody, biotin conjugate validated for use in ELISA, FC, WB, IC, IH(P).
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Target description

25 kDa

Physical form

ImmunoAffinity Purified
Lyophilized powder from a solution with 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15 mg/mL BSA as stabilizer. Purified by immunoaffinity chromatography.

Storage and Stability

Maintain lyophilized product at 2°-8°C for up to 12 months. After reconstitution the product is stable for six weeks at 2°-8°C in the dark as an undiluted liquid. Reconstitute vial with of distilled water (addition of 0.5 ml water will yield a final concentration of 1 mg/mL). For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze thaw cycles. Keep away from light.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical implementation, characterization and applications of a multi-colour time-gated luminescence microscope.
Zhang, L; Zheng, X; Deng, W; Lu, Y; Lechevallier, S; Ye, Z; Goldys, EM; Dawes, JM; Piper et al.
Scientific Reports null
Dayong Jin et al.
Journal of biomedical optics, 14(2), 024023-024023 (2009-05-02)
A fundamental problem for rare-event cell analysis is auto-fluorescence from nontarget particles and cells. Time-gated flow cytometry is based on the temporal-domain discrimination of long-lifetime (>1 micros) luminescence-stained cells and can render invisible all nontarget cell and particles. We aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service