Skip to Content
Merck
All Photos(1)

Key Documents

AP130C

Sigma-Aldrich

Goat Anti-Mouse IgG & IgM Antibody, Cy3 conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$486.00

$486.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
2 MG
$486.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$486.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

This Goat anti-Mouse IgG & IgM Antibody, Cy3 conjugate is validated for use in IF for the detection of Mouse IgG & IgM.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quentin Delmotte et al.
Frontiers in cellular neuroscience, 14, 98-98 (2020-05-20)
Sonic hedgehog (Shh) signaling plays critical roles during early central nervous system development, such as neural cell proliferation, patterning of the neural tube and neuronal differentiation. While Shh signaling is still present in the postnatal brain, the roles it may
Yuting Xiang et al.
Microsystems & nanoengineering, 8, 70-70 (2022-07-02)
Electrophysiological recording is a widely used method to investigate cardiovascular pathology, pharmacology and developmental biology. Microelectrode arrays record the electrical potential of cells in a minimally invasive and high-throughput way. However, commonly used microelectrode arrays primarily employ planar microelectrodes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service