Skip to Content
Merck
All Photos(1)

Key Documents

AG4000

Sigma-Aldrich

uPA Colorimetric Substrate, ECM600, ECM610

Synonym(s):

ECM 600 substrate, ECM 610 substrate, uPA chromogenic substrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$329.00

$329.00


Estimated to ship on18 May 2025



Select a Size

Change View
5 MG
$329.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

$329.00


Estimated to ship on18 May 2025


product line

Chemicon®

Quality Level

technique(s)

activity assay: suitable

General description

pyroGLU-GLY-ARG-pNA-HCL {L-Pyroglutamyl-glycyl-L-arginine-p-Nitroaniline hydrochloride

Mol wt. 498.9
Ex (316nm) 1.27x10(e-1)/mol/cm

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service