Skip to Content
Merck
All Photos(1)

Key Documents

ABT267

Sigma-Aldrich

Anti-CELSR3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cadherin EGF LAG seven-pass G-type receptor 3, Cadherin family member 11, Epidermal growth factor-like protein 1, EGF-like protein 1, Flamingo homolog 1, hFmi1, Multiple epidermal growth factor-like domains protein 2,

Short name=Multiple EGF-li

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$404.00

$404.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$404.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$404.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CELSR3(1951)

General description

CELSR3, also known as Cadherin family member 11, Epidermal growth factor-like protein 1, EGF-like protein 1 or Flamingo homolog 1, FML1, or Multiple epidermal growth factor like domains protein 2, and encoded by the gene CELSR3/CDHF11, EGFL1, FMI1, KIAA0812, MEGF2, is a cell surface receptor important for cell to cell signaling and positioning particularly during the development of the nervous system. It is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined but may be involved in the regulation of contact-dependent neurite growth and may play a role in tumor formation, and recent research demonstrates that CELSR3 is critical for proper development of GABA-signaling pathways in the inner retina. As a cadherin member, CELSR3 is localized to the cell membrane.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain within the C-terminus of human CELSR3.

Application

Detect CELSR3 using this rabbit polyclonal antibody, Anti-CELSR3 Antibody validated for use in IHC.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Immunohistochemistry in human cerebellum and glioma sections.

Immunohistochemistry Analysis: A 1:2000 dilution and a 1:500 dilution of this antibody detected CELSR3 in human cerebellum and glioma sections, respectively.

Target description

358 kDa calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service