Skip to Content
Merck
All Photos(2)

Key Documents

ABT22

Sigma-Aldrich

Anti-Endophilin-A1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SH3-domain GRB2-like 2, bA335L15.1 (SH3-domain GRB2-like 2), Endophilin A1 BAR domain, SH3 domain protein 2A, Endophilin-1, SH3 domain-containing GRB2-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$656.00

$656.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$656.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$656.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Sh3Gl2(20404)

General description

A member of the Endophilin family, Endophilin A1 is a protein containing SH3 domains, and has been implicated in the trafficking of clathrin-mediated synaptic vesicle endocytosis within the membrane. It was originally characterized as a presynaptically enriched protein that binds to endocytic proteins polyphosphoinositide synaptojanin and dynamin. Endophilin A1 has been implicated in endocytic vesicle budding when its function in cell-free systems and in living synapses has been disturbed. Along with the SH3 domain, Endophilin A1 contains a BAR domain in the N-terminal region, that has been found to induce and bind membrane curvature. This protein’s expression is localized to the brain, most specifically in the frontal cortex. Markedly high expression is observed in fetal cerebellum.

Specificity

Human (94% sequence homology).
The antibody recognizes Endophilin-A1.

Immunogen

KLH-conjugated linear peptide corresponding to mouse Endophilin-A1.

Application

This Anti-Endophilin-A1 Antibody is validated for use in WB for the detection of Endophilin-A1.
Western Blot Analysis: 1:500 dilution from a previous lot detected Endophilin-A1 in 10 µg of rat brain and mouse brain membrane tissue lysates.

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1:500 dilution of this antibody detected Endophilin-A1 in 10 µg of mouse brain tissue lysate.

Target description

~ 42 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service