Skip to Content
Merck
All Photos(1)

Key Documents

ABT142

Sigma-Aldrich

Anti-PLK-4 Antibody

from rabbit

Synonym(s):

Serine/threonine-protein kinase PLK4, Polo-like kinase 4, PLK-4, Serine/threonine-protein kinase 18, Serine/threonine-protein kinase Sak

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$750.00

$750.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$750.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$750.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLK4(10733)

General description

Serine/threonine-protein kinase PLK4 (PLK-4) is a member of the protein kinase superfamily and plays an essential role in centriole duplication. PLK-4 triggers procentriole formation on the surface of the parental centriole cylinder, leading to the recruitment of centriole biogenesis proteins. It may be involved in tumorigenesis since centrosome aberrations are frequently observed in tumors. PLK-4 may also be involved in trophoblast differentiation by phosphorylating HAND1, leading to the disruption between HAND1 and MDFIC and then activating HAND1.

Specificity

This antibody recognizes the POLO box domain of PLK-4

Immunogen

Epitope: POLO box domain
KLH-conjugated linear peptide corresponding to the POLO box domain of human PLK-4.

Application

Anti-PLK-4 Antibody detects level of PLK-4 & has been published & validated for use in Western Blotting, ICC.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected PLK-4 in 10 µg of Jurkat cell lysate.

Target description

~95 kDa observed

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service