Skip to Content
Merck
All Photos(3)

Key Documents

ABT119

Sigma-Aldrich

Anti-CELSR1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cadherin EGF LAG seven-pass G-type receptor 1, Cadherin family member 9, Flamingo homolog 2, hFmi2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$673.00

$673.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$673.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$673.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CELSR1(9620)

General description

The Flamingo/CELSR family of 7TM-cadherins are characterized with a symmetric protein distribution and polarized activity at neighboring epithelial cell interfaces along defined axes of planar cell polarity. As a component of the extracellular matrix, the CELSR1 protein has been found particularly at the basal surface of neuroepithelial cells within both the early neural tube and a less well-defined group of ventricular zone cells at the midline of the developing spinal cord. CELSR1 plays a role in orienting hair follicles along the anterior–posterior axis of the developing mouse epidermis, both during embryogenesis and postnatal development.

Immunogen

Linear peptide corresponding to human CELSR1.

Application

Immunofluorescence Analysis: A 1:500 and 1:1,000 dilution from a representative lot detected CELSR1 in rat cerebellum, substancia nigra, and hippocampus tissue.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected CELSR1 in neurons of rat cortex tissue and in Purkinje cells and cells within the granular layer of rat cerebellum tissue.
This Anti-CELSR1 Antibody is validated for use in Western Blotting, IHC, Immunofluorescence for the detection of CELSR1.

Quality

Evaluated by Western Blot in PC12 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected CELSR1 in 10 µg of PC12 cell lysate.

Target description

~330 kDa observed. A second isoform at ~152 kDa may be observed in some lysates.
An uncharacterized band at ~54 kDa may be observed in some cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengwen An et al.
Nature communications, 9(1), 378-378 (2018-01-27)
The extent to which heterogeneity within mesenchymal stem cell (MSC) populations is related to function is not understood. Using the archetypal MSC in vitro surface marker, CD90/Thy1, here we show that 30% of the MSCs in the continuously growing mouse
Lena P Basta et al.
Frontiers in cell and developmental biology, 10, 1064907-1064907 (2023-01-31)
Cadherin EGF LAG seven-pass G-type receptor (Celsr) proteins 1-3 comprise a subgroup of adhesion GPCRs whose functions range from planar cell polarity (PCP) signaling to axon pathfinding and ciliogenesis. Like its Drosophila ortholog, Flamingo, mammalian Celsr1 is a core component

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service