Skip to Content
Merck
All Photos(2)

Key Documents

ABT110

Sigma-Aldrich

Anti-SDHD (CybS) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial, CybS, CII-4, QPs3, Succinate dehydrogenase complex subunit D, Succinate-ubiquinone oxidoreductase cytochrome b small subunit, Succinate-ubiquinone reductase membrane anchor s

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$750.00

$750.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$750.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$750.00


Estimated to ship on13 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

sheep (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SDHD(6392)

General description

Succinate dehydrogenase complex subunit D (SDHD) is a member of the CybS family and is one of four subunits of the succinate dehydrogenase (SDH) complex involved in the electron transport chain of the mitochondria. The specific role of SDH is to transfer electrons from succinate to ubiquinone, and the SDHD subunit is involved in anchoring the SDH complex to the inner mitochondrial membrane. Defective SDHD has been linked to Carney-Stratakis syndrome, a condition characterized by concurrent paraganglioma and gastric stromal sarcomas.

Specificity

Other homologies: Rat (92% sequence homology).
This antibody recognizes the Topological domain (mitochondrial matrix) of SDHD (CybS).

Immunogen

Epitope: Topological domain (mitochondrial matrix)
KLH-conjugated linear peptide corresponding to the Topological domain of human SDHD (CybS).

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected SDHD (CybS) in NIH/3T3 and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-SDHD (CybS) Antibody is validated for use in Western Blotting, ICC for the detection of SDHD (CybS).

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected SDHD (CybS) in 10 µg of HeLa cell lysate.

Target description

~11 kDa observed. An uncharacterized band may be observed at ~25 kDa in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte L Alston et al.
Human genetics, 134(8), 869-879 (2015-05-27)
Succinate dehydrogenase (SDH) is a crucial metabolic enzyme complex that is involved in ATP production, playing roles in both the tricarboxylic cycle and the mitochondrial respiratory chain (complex II). Isolated complex II deficiency is one of the rarest oxidative phosphorylation
Yoshihiro Hayashi et al.
Cancer discovery, 8(11), 1438-1457 (2018-08-25)
Myelodysplastic syndromes (MDS) are heterogeneous hematopoietic disorders that are incurable with conventional therapy. Their incidence is increasing with global population aging. Although many genetic, epigenetic, splicing, and metabolic aberrations have been identified in patients with MDS, their clinical features are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service