Skip to Content
Merck
All Photos(2)

Key Documents

ABS988

Sigma-Aldrich

Anti-ACAT-2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SOAT2_HUMAN, SOAT2, ACACT2, ACTL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$698.00

$698.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$698.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$698.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, monkey

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACAT2(39)

General description

Acyl-coenzyme A:cholesterol acyltransferase 2 (ACAT-2), also known as Sterol O-acyltransferase 2
(SOAT2), is a synthetic enzyme that is important in lipoprotein assembly, steroid metabolism, and dietary cholesterol metabolism and absorption. ACAT-2 is expressed primarily in the liver and intestine in enterocytes in their endoplasmic reticulum. ACAT-2 esterifies cholesterol allowing it to be more efficiently packaged and transported. Free cholesterol can be taken up by lipoproteins but it is confined to the outer surface of the lipoparticle. Esterification allows internal packaging and much more efficient transport through the blood stream. Gene expression of ACAT-2 is partially regulated by the transcriptional repressor TGIF1 as well as other factors such as thyroid hormone expression and microRNA miR181d.

Immunogen

Recombinant protein corresponding to Human ACAT-2.

Application

Immunocytochemistry Analysis: A representative lot detected ACAT-2 in human liver tissue (Parini, P., et al. (2004). Circulation. 2004;110:2017-2023).
Western Blot Analysis: A representative lot detected ACAT-2 in human liver lysate (Parini, P., et al. (2004). Circulation. 2004;110:2017-2023)
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation
This Anti-ACAT-2 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of ACAT-2.

Quality

Evaluated by Western Blotting in human lung tissue lysate using a non-reduced gel.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ACAT-2 in 10 µg of human lung tissue lysate using a non-reduced gel

Target description

~ 55 kDa observed, uncharacterized bands may appear at lower molecular weight

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 20 mM NaPO4, 28 mM NaCl and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service