Skip to Content
Merck
All Photos(3)

Documents

ABS82-AF488

Sigma-Aldrich

Anti-VEGF Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Vascular endothelial growth factor A, VEGF-A, Vascular permeability factor, VPF, VEGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VEGFA(7422)

General description

VEGF (Vascular endothelial growth factor, VEGFA, VEGF-A), a dimeric ligand, is among the most potent angiogenic factor. VEGF is secreted by tumor cells and other cells exposed to hypoxia. VEGF is a highly specific mitogen for vascular endothelial cells. Seven VEGF isoforms (a, b, c, d, e, f, g) are generated as a result of alternative splicing from a single VEGF gene. The most commonly studied isoforms are VEGF121, VEGF165, and VEGF189 (f, d and b, respectively) All seven isoforms differ in their molecular mass and in biological properties, such as their ability to bind to cell-surface heparan-sulfate proteoglycans. The expression of VEGF is potentiated in response to hypoxia, by activated oncogenes, and by a variety of cytokines. VEGF also enhances permeabilization of blood vessels and plays a central role in the regulation of vasculogenesis. VEGF expression has been linked to the degree of tumor vascularization and with increased metastatic risk. Additionally, VEGF induces endothelial cell proliferation, promotes cell migration, and inhibits apoptosis. Consequently, inhibition of VEGF signaling abrogates the development of a wide variety of tumors.

Specificity

Other homologies: Bovine (85% sequence homology). Rat (70% sequence homology). Mouse (65% sequence homology).
This antibody recognizes VEGF at the N-terminus.

Immunogen

Epitope: N-terminus.
KLH-conjugated linear peptide corresponding to human VEGF at the N-terminus.

Application

Anti-VEGF Antibody, Alexa Fluor 488 Conjugate is an antibody against VEGF for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected VEGF in HeLa and A431 cells.
The unconjugated antibody (Cat. No. ABS82) is shown to be suitable also for Western blotting and immunohistochemistry (Paraffin) applications.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Evaluated by Immunocytochemistry in NIH/3T3 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected VEGF in NIH/3T3 cells.

Target description

~55 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody conjugate in PBS with 15mg/ml BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service