Skip to Content
Merck
All Photos(1)

Key Documents

ABS763

Sigma-Aldrich

Anti-DPAGT1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

UDP-N-acetylglucosamine--dolichyl-phosphate N-acetylglucosaminephosphotransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$576.00

$576.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$576.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$576.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DPAGT1(1798)

General description

UDP-N-acetylglucosamine-dolichyl-phosphate N-acetylglucosaminephosphotransferase, also known as GlcNAc-1-P transferase (G1PT or GPT) or N-acetylglucosamine-1-phosphate transferase, and encoded by the gene name DPAGT1 or DPAGT2, is an enzyme that catalyzes the first step in the dolichol-linked oligosaccharide pathway for glycoprotein biosynthesis. Mutations in this integral endoplasmic reticulum (ER) membrane protein enzyme belongs to the glycosyltransferase family 4 results in the congenital disorder of glycosylation type Ij with symptoms such as severe hypotonia, medically intractable seizures, mental retardation, microcephaly, and exotropia. Recent experiments have shown that DPAGT1 is a target of the Wnt/beta-catenin signaling pathway, with Wnt3a inducing higher DPAGT1 mRNA expression.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of human DPAGT1.

Application

Anti-DPAGT1 Antibody detects level of DPAGT1 & has been published & validated for use in DPAGT1.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse kidney tissue lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected DPAGT1 in mouse kidney tissue lysate with and without blocking peptide.

Target description

~42 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service