Skip to Content
Merck
All Photos(2)

Key Documents

ABS523

Sigma-Aldrich

Anti-Lipin-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidate phosphatase LPIN1, Lipin-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$467.00

$467.00


Estimated to ship on19 May 2025



Select a Size

Change View
100 μG
$467.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$467.00


Estimated to ship on19 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LPIN1(23175)

General description

Lipin-1 is also called Phosphatidate phosphatase LPIN1. Lipin-1 controls the metabolism of fatty acids, acts as a magnesium-dependent phosphatidate phosphatase enzyme, as well as a nuclear transcriptional coactivator that modulates lipid metabolism gene expression. Lipin-1 involved in adipocyte differentiation, and is also potentially involved in mitochondrial fission. Lipin-1 is specifically expressed in skeletal muscle, and is also abundant in adipose tissue. Lipin-1 defects can cause autosomal recessive acute recurrent myoglobinuria (ARARM).

Immunogen

linear peptide corresponding to human Lipin-1.

Application

Anti-Lipin-1 Antibody is an antibody against Lipin-1 for use in IHC, western blotting & IP.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Lipin-1 in human cardiac myocytes and human skeletal muscle myocytes tissue.
Western Blotting Analysis: A representative lot detected Lipin-1 in mouse adipose tissue lysate (Mayurranjan, S., et al. (2013). PNAS. 110(2):642-647).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Lipin-1 in adipose tissue lysate (Mayurranjan, S., et al. (2013). PNAS. 110(2):642-647).
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Evaluated by Western Blotting in mouse hepatocytes overexpressing lipin1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Lipin-1 in 10 µg of mouse hepatocytes overexpressing lipin1 cell lysate (lysate from the laboratory of Brian Finck, Washington University, St. Louis).

Target description

~ 110 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide with 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service