Skip to Content
Merck
All Photos(2)

Key Documents

ABS518

Sigma-Aldrich

Anti-SLC5A3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Sodium/myo-inositol cotransporter, Na(+)/myo-inositol cotransporter, Sodium/myo-inositol transporter 1, SMIT1, Solute carrier family 5 member 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$642.00

$642.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$642.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$642.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC5A3(6526)

General description

SLC5A3, also known as Sodium/myo-inositol cotransporter, Na (+)/myo-inositol cotransporter, Sodium/myo-inositol transporter 1 (SMT1), or Solute carrier family 5 member 3, and encoded by the gene SLC5A3, is an important intracellular transporter and signaling mediator that mediates the intracellular accumulation of myo-inositol. The compound myo-inositol is important for cell signaling both in the cytoplasm and in intracellular organelles. It is required in the plasma membrane and in the cytoplasm for maintaining the synthesis of the second messengers such as Inositol trisphosphate (IP3) and diacylglycerol (DAG) from phosphatidylinositol bisphosphate (PIP2). In organelles myo-inositol is a precursor for the synthesis of complex signaling phospholipids and inositolphosphates from IP3 and PIP2 as well. Moreover, SLC5A3 is upregulated by osmotic shock and downregulated by energy depletion and serves as a molecular sensor of cellular stress.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human SLC5A3.

Application

Detect SLC5A3 using this Anti-SLC5A3 Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected SLC5A3 in human prostate tissues.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected SLC5A3 in human placenta tissues.

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected SLC5A3 in 10 μg of HUVEC cell lysate.

Target description

~79 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Zheng et al.
Cell reports. Medicine, 4(6), 101057-101057 (2023-06-02)
Single-cell transcriptomics can provide quantitative molecular signatures for large, unbiased samples of the diverse cell types in the brain. With the advances of multi-omics datasets, a major challenge is to validate and integrate results into a biological understanding of spatial
Heidy Cabrera-Cruz et al.
American journal of physiology. Endocrinology and metabolism, 318(2), E237-E248 (2019-12-25)
Polycystic ovary syndrome (PCOS) is an endocrine-metabolic disorder characterized by hyperandrogenism and ovulatory dysfunction but also obesity and hyperinsulinemia. These characteristics induce an insulin-resistant state in tissues such as the endometrium, affecting its reproductive functions. Myo-inositol (MYO) is an insulin-sensitizing
April R Foster et al.
Aging cell, 19(1), e13058-e13058 (2019-11-27)
Aging is characterized by the deterioration of tissue structure and function. In skin, environmental factors, for example, ultraviolet radiation (UVR), can accelerate the effects of aging such as decline in barrier function and subsequent loss of hydration. Water homeostasis is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service