Skip to Content
Merck
All Photos(2)

Key Documents

ABS492

Sigma-Aldrich

Anti-RhoGAP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Rho GTPase-activating protein 1, CDC42 GTPase-activating protein, GTPase-activating protein rhoOGAP, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1, p50-RhoGAP, RhoGAP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$416.00

$416.00


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$416.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$416.00


Estimated to ship on09 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARHGAP1(392)

General description

RhoGAP1, also known as Rho GTPase-activating protein 1, CDC42 GTPase-activating protein, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1, or p50-RhoGAP, and encoded by the gene ARHGAP1/CDC42GAP/RHOGAP1, is an important GTPase activator for the Rho, Rac, and CDC42 GTP binding proteins, converting them to the putatively inactive GDP-bound state. CDC42 is the preferred substrate for RhoGAP1. Such GTPase-activating proteins or GAPs play an important role in signal transduction pathways regulated by GTP-binding proteins. In addition to acting as down-regulators of GTPases, there is growing evidence that they also act as effector molecules required for downstream signaling. RhoGAP1 is found in multiprotein complexes in the cytoplasm in most cells. Recent research has shown RhoGAP1 to be particularly important during epithelial to mesenchymal morphogenesis and for carcinoma metastasis. Rho-ROCK signaling is essential for cell apical detachment and the generation of motility. RhoGAP1 restricts Rho activation to apical areas and thus helps guide the detachment process and thus is a target of much developmental and cancer research.

Immunogen

GST-tagged recombinant protein corresponding to human RhoGAP1.

Application

Anti-RhoGAP1 Antibody is an antibody against RhoGAP1 for use in western blotting & ICC.
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected RhoGAP1 in 10 µg of A431, NIH/3T3, and PC-12 cell lysates.

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected RhoGAP1 in HeLa cells (Schiel, J. A., et al. (2012). Nat Cell Biol. 14(10):1068-1078.).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected RhoGAP1 in 10 µg of HeLa cell lysate.

Target description

~50 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service