Skip to Content
Merck
All Photos(2)

Key Documents

ABS232

Sigma-Aldrich

Anti-MIXL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Homeobox protein MIXL1, Homeodomain protein MIX, hMix, MIX1 homeobox-like protein 1, Mix.1 homeobox-like protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$610.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MIXL1(83881)

General description

MIXL1 is also known as Homeobox protein MIXL1, Homeodomain protein MIX (hMix), MIX1 homeobox-like protein 1, and Mix.1 homeobox-like protein. MIXL1 is a transcription factor with roles in axial mesendoderm morphogenesis, endoderm formation, blood cell differentiation, and morphogenesis of the heart and the gut during embryogenesis. MIXL1 has been shown to negatively regulate brachyury expression. MIXL1 is expressed mainly in progenitors and secondary lymph tissues, immature B- and T-lymphoid cells during normal hematopoiesis, and is also present in differentiating embryonic stem cells.

Specificity

Other homologies: Mouse and Rat (85% sequence homology).
This antibody recognizes the homeobox DNA binding domain of MIXL1.

Immunogen

Epitope: Homeobox DNA binding domain
KLH-conjugated linear peptide corresponding to the homeobox DNA binding domain of human MIXL1.

Application

Anti-MIXL1 Antibody is a highly specific rabbit polyclonal antibody, that targets Homeobox protein MIXL1 & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected MIXL1 in H9 embryoid body cells. This protein was observed in H9 embryoid bodies (Lee, M. R., et al. (2010). Stem Cells. 28:1550-1559). MIXL1 was also localized in the nucleus of COS-7 and mouse ES cells (Mossman, A. K., et al. (2005). Stem Cells and Development. 14:656-663).
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MIXL1 in 10 µg of Jurkat cell lysate.

Target description

~36 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarel K Gandhi et al.
Stem cells translational medicine, 8(6), 512-521 (2019-02-16)
Human fibrin hydrogels are a popular choice for use as a biomaterial within tissue engineered constructs because they are biocompatible, nonxenogenic, autologous use compatible, and biodegradable. We have recently demonstrated the ability to culture induced pluripotent stem cell (iPSC)-derived retinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service