Skip to Content
Merck
All Photos(1)

Key Documents

ABS1491

Sigma-Aldrich

Anti-Frizzled-2

from rabbit, purified by affinity chromatography

Synonym(s):

Fz-2, hFz2, FzE2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$604.00

$214.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$604.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FZD2(2535)

General description

Frizzled-2 (UniProt: Q14332; also known as Fz-2, hFz2, FzE2) is encoded by the FZD2 gene (Gene ID: 2535) in human. Frizzled-2 is a multi-pass membrane protein that is widely expressed in adult tissues. In fetus, higher levels are expressed in the brain, lung, and kidney and lower levels are observed in liver. Frizzled serves as a receptor for Wnt proteins. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. In the presence of Wnt the Frizzled receptor is activated, which along with low-density lipoprotein receptor-related proteins (LRP) 5 and 6 stabilizes beta-catenin. The formation of Wnt-Frizzled-LRP6 complex, together with the recruitment of the scaffolding protein Dishevelled (Dvl), results in LRP6 phosphorylation and activation and the recruitment of the Axin complex to the receptors. This leads to the inhibition of Axin-mediated beta-catenin phosphorylation, which stabilizes beta-catenin. Frizzled-2 is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off in the mature form. Its Fz domain is localized to amino acids 34-153 and this domain is involved in its binding with Wnt ligands. Frizzled-2 can be ubiquitinated by RING-type E3 ubiquitin transferase ZNRF3 leading to its proteasomal degradation. Abnormalities in Wnt signaling are reported to promote birth defects, degenerative diseases, and cancer.

Specificity

This rabbit polyclonal antibody specifically detects human Frizzled -2. It targets an epitope with in 13 amino acids from the extracellular domain.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal half of human Frizzled-2.

Application

Detects Frizzled-2 using this rabbit polyclonal Anti-Frizzled-2 Antibody, Cat. No. ABS1491, tested for use in Western Blotting.
Research Category
Signaling

Quality

Evaluated by Western Blotting in CACO-2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Frizzled-2 in CACO-2 cell lysate.

Target description

~60 kDa observed; 63.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service