Skip to Content
Merck
All Photos(3)

Key Documents

ABS1035

Sigma-Aldrich

Anti-FMO2 Antibody

serum, from rabbit

Synonym(s):

Dimethylaniline monooxygenase [N-oxide-forming] 2, Dimethylaniline oxidase 2, FMO 1B1, Pulmonary flavin-containing monooxygenase 2, FMO 2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$479.00

$479.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$479.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$479.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), orangutan (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FMO2(2327)

General description

Dimethylaniline monooxygenase [N-oxide-forming] 2 (EC 1.14.13.8; UniProt Q99518; also known as Dimethylaniline oxidase 2, FMO 1B1, FMO 2, Pulmonary flavin-containing monooxygenase 2) is encoded by the FMO2 (also known as FMO1B1) gene (Gene ID 2327) in human. Flavin monooxygenases (FMOs) are flavin-containing enzymes that catalyze substrates oxidation at the expense of NADPH and molecular oxygen. FMO2 is expressed in the lung tissue and therefore known as the pulmonary FMO. Two FMO2 alleles have been described in human. FMO2.1 is the functional full-length version that is found in high frequency in Sub-Saharan African populations (49%) and their Hispanic descendants (2-7%), whereas the FMO2.2 gene carries a C-to-T transition mutation that results in a premature stop codon (TAG), yielding a C-terminally truncated 64-amino acid non-functional protein FMO2X472 found in all Caucasians and Asians. Known human FMO2.1 substrates include sulfur-derived compounds such as thioureas, thioetherorganophosphates, thiazetazone and ethionamide.

Immunogen

Epitope: Internal (N-terminal half).
Linear peptide corresponding to an internal sequence from the N-terminal half of human FMO2.

Application

Detect FMO2 using this Anti-FMO2 Antibody validated for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected FMO2 in mouse lung, mouse kidney, and human kidney tissue sections.
Research Category
Signaling
Research Sub Category
Toxicology & Drug Resistance

Quality

Evaluated by Western Blotting in human liver microsomal lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected FMO2 in 10 µg of human liver microsomal lysate.

Target description

~55 kDa observed. 53.64 kDa calculated.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service