Skip to Content
Merck
All Photos(2)

Key Documents

ABS1009

Sigma-Aldrich

Anti-HSD17B7 Antibody

Synonym(s):

3-keto-steroid reductase|17-beta-hydroxysteroid dehydrogenase 7|17-beta-HSD 7|Estradiol 17-beta-dehydrogenase 7|PRL receptor-associated protein|PRAP|HSD17B7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

human, hamster, mouse, rat, bovine

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

General description

3-keto-steroid reductase (UniProt Q62904; EC 1.1.1.270, EC 1.1.1.62; also known as PRAP, 17-beta-HSD 7, PRL receptor-associated protein, Estradiol 17-beta-dehydrogenase 7, 17-beta-hydroxysteroid dehydrogenase 7, Hydroxysteroid dehydrogenase 17 beta, type 7) is encoded by the Hsd17b7 gene in rat (Entrez Gene ID 29540). Hsd17b7 protein is most abundantly expressed in ovaries of pregnant animals and functions both as a 17-beta-hydroxysteroid dehydrogenase (EC 1.1.1.62) in the biosynthesis of sex steroids, as well as as a 3-ketosteroid reductase (EC 1.1.1.270) in the biosynthesis of cholesterol. Cat. No. ABS1009, Anti-HSD17B7, is a rabbit polyclonal antiserum raised against native rat protein, and shows reactivity toward rat, human, mouse, hamster, and bovine species (PMID 1425419).

Immunogen

Purified protein corresponding to rat HSD17B7.

Application

Anti-HSD17B7 Antibody is an antibody against HSD17B7 for use in Western Blotting, Immunohistochemistry, Immunocytochemistry.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected HSD17B7 in mouse ovary sections and human adrenal gland tissue.
Western Blotting Analysis: A representative lot detected HSD17B7 in rat, mouse, cow, hamster, bovine,

Quality

Evaluated by Western Blotting in mouse stomach tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSD17B7 in 10 ug of mouse stomach tissue lysate.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service