Skip to Content
Merck
All Photos(2)

Key Documents

ABN68

Sigma-Aldrich

Anti-Bystin Antibody

from rabbit, purified by affinity chromatography

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$383.00

$383.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$383.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$383.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, horse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), equine (based on 100% sequence homology), canine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BYSL(705)

General description

Bystin and tastin serve as cytoplasmic proteins that are required for trophinin to mediate cell-to-cell adhesion between trophoblastic and endometrial epithelial cells in human placenta. Although the trophinin gene is unique to mammals, the bystin gene (BYSL) is conserved across eukaryotes. It is shown that bystin influences the transition from silent trophectoderm to an active trophoblast through trophinin-mediated cell adhesion. It is also essential for embryonic stem cell survival and trophectoderm development in mouse. Bystin is demonstrated to play a key role in ribosomal biogenesis by processing 18S RNA in the 40S subunit in human cancer cells and mouse embryos. It is also reported that c-MYC targets BYSL, which corresponds with bystin-mediated, rapid protein synthesis, and subsequent cellular growth.

Specificity

This antibody recognizes the C-terminus of Bystin.

Immunogen

Epitope: C-terminus.
KLH-conjugated linear peptide corresponding to the C-terminus of human Bystin.

Application

Anti-Bystin Antibody is an antibody against Bystin for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:300 dilution of this antibody detected Bystin in the syncytiotrophoblastic layer of the chorionic villi in human placenta.
Research Category
Neuroscience
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Bystin in 10 µg HeLa cell lysate.

Target description

~55 kDa observed. UniProtKB/Swiss-Prot Q13895 (BYST_HUMAN) states a molecular weight of ~49 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service