Skip to Content
Merck
All Photos(2)

Key Documents

ABN478

Sigma-Aldrich

Anti-LFG Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein lifeguard 2, Fas apoptotic inhibitory molecule 2, Neural membrane protein 35, Transmembrane BAX inhibitor motif-containing protein 2, LFG

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$491.00

$491.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$491.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$491.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FAIM2(23017)

General description

Protein lifeguard 2 (LFG) is an antiapoptotic protein belonging to the BI1 family. LFG inhibits the apoptotic signal transduced by the Fas receptor but not from the related tumor necrosis factor-death signal. LFG is expressed at high levels in the brain especially in the hippocampus. It is also highly expressed in breast cancer tissues and correlates with the grade of tumor (grade II/grade III) in primary breast tumors.

Immunogen

KLH-conjugated linear peptide corresponding to human LFG.

Application

Detect LFG using this rabbit polyclonal antibody, Anti-LFG Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected LFG in mouse brain tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in EL4 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LFG in 15 µg of EL4 cell lysate.

Target description

~35 kDa observed.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
EL4 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service