Skip to Content
Merck
All Photos(5)

Key Documents

ABN44

Sigma-Aldrich

Anti-FGF10 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fibroblast growth factor 10, FGF-10, Keratinocyte growth factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$506.00

$506.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$506.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$506.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FGF10(2255)

General description

Fibroblast growth factor 10 (FGF10) is a member of the fibroblast growth factor family and plays an essential role in the regulation of cell differentiation, tissue repair, tumor growth, embryonic development, and cell proliferation. Mutations in FGF10 have been associated lacrimo-auriculo-dento-digital syndrome (LADDS) which is a form of ectodermal dysplasia.

Immunogen

KLH-conjugated linear peptide corresponding to human FGF10.

Application

Detect FGF10 using this Anti-FGF10 Antibody validated for use in WB, IH(P).
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blot Analysis: 0.5 µg/mL from a representative lot detected FGF10 on 10 µg of mouse embryo tissue lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected FGF10 in mouse epithelial cells, mouse liver, human brain, and human cerebellum tissues.

Quality

Evaluated by Western Blot in A549 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected FGF10 on 10 µg of A549 cell lysate.

Target description

~23 kDa observed

Physical form

Affinity purified
Purified Rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Wang et al.
Cell proliferation, 55(11), e13315-e13315 (2022-07-20)
Ultraviolet light B (UVB) irradiation can induce skin injury and result in keratinocytes proliferation inhibition. However, the molecular understanding of the repair during UVB-induced cell proliferation inhibition remains poorly understood. The purpose of this study was to explore the role
Shawn Liang et al.
Development (Cambridge, England), 145(2) (2018-01-24)
The developmental program that regulates thyroid progenitor cell proliferation is largely unknown. Here, we show that branching-like morphogenesis is a driving force to attain final size of the embryonic thyroid gland in mice. Sox9, a key factor in branching organ
Roberta Haddad-Tóvolli et al.
American journal of physiology. Endocrinology and metabolism, 324(2), E154-E166 (2023-01-05)
Maternal obesity is an important risk factor for obesity, cardiovascular, and metabolic diseases in the offspring. Studies have shown that it leads to hypothalamic inflammation in the progeny, affecting the function of neurons regulating food intake and energy expenditure. In
Grinu Mathew et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(46), 13156-13161 (2016-11-02)
Deficiency in PTEN (phosphatase and tensin homolog deleted on chromosome 10) is the underlying cause of PTEN hamartoma tumor syndrome and a wide variety of human cancers. In skin epidermis, we have previously identified an autocrine FGF signaling induced by
Thomas Volckaert et al.
Developmental cell, 43(1), 48-59 (2017-10-11)
The lung harbors its basal stem/progenitor cells (BSCs) in the protected environment of the cartilaginous airways. After major lung injuries, BSCs are activated and recruited to sites of injury. Here, we show that during homeostasis, BSCs in cartilaginous airways maintain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service