Skip to Content
Merck
All Photos(2)

Key Documents

ABN426

Sigma-Aldrich

Anti-phospho-Neurogranin (Ser36)/Neuromodulin (Ser41) Antibody

serum, from rabbit

Synonym(s):

Protein kinase C substrate 7.5 kDa protein RC3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$705.00

$705.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$705.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$705.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer36)

Gene Information

mouse ... Nrgn(64011)
rat ... Nrgn(64356)

General description

Neurogranin (Ng) (also named RC3, p17 or BICKS) is a small protein originally identified in rat brain and abundantly expressed in several telencephalic areas, such as the cerebral cortex, hippocampus, amygdala, and striatum. In neurons, it is found concentrated at dendritic spines where it participates in synaptic signaling events through the regulation of calmodulin (CaM). Neurogranin protein is a critical third messenger and substrate of PKC in the molecular cascade necessary for synaptic development and remodeling. Neurogranin (Ng) features an IQ motif that mediates its interaction with CaM and phosphatidic acid (PA). The interaction is controlled by phosphorylation at serine 36 of Neurogranin. Neurogranin is phosphorylated at serine 36 by PKC. Ser36-phosphorylated Neurogranin is unable to bind either CaM or PA. Neurogranin in localized in the cell bodies of neurons in the cortex and in the apical and basal dendrites of pyramidal neurons. Neurogranin is not found in dendrites and its expression is very low as well in Alzheimer’s disease patients.

Specificity

Phosphorylated Neurogranin and Neuromodulin

Immunogen

Epitope: IQ domain
KLH-conjugated linear peptide corresponding to the IQ domain of Rat phospho-Neurogranin.

Application

Research Category
Neuroscience
Research Sub Category
Neuroregenerative Medicine
This Anti-phospho-Neurogranin (Ser36)/Neuromodulin (Ser41) Antibody is validated for use in Western Blotting for the detection of phospho-Neurogranin (Ser36)/Neuromodulin (Ser41).

Quality

Evaluated by Western Blotting in Mouse brain tissue lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected phospho-Neurogranin (Ser36) / Neuromodulin (Ser41) in 10 µg of Mouse brain tissue lysate treate with magnisium (lane1) or PKC (lane2)

Target description

~43 kDa is phospho-Neuromodulin (GAP-43) and ~17 KD is phospho-Neurogranin. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit Polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Svirsky et al.
Molecular neurobiology, 61(9), 7256-7268 (2024-02-20)
Extensive effort has been made to study the role of synaptic deficits in cognitive impairment after traumatic brain injury (TBI). Neurogranin (Ng) is a calcium-sensitive calmodulin (CaM)-binding protein essential for Ca2+/CaM-dependent kinase II (CaMKII) autophosphorylation which subsequently modulates synaptic plasticity.
Shu Xu et al.
Frontiers in psychiatry, 12, 763032-763032 (2021-11-16)
Objective: Rapid eye movement sleep deprivation (REM-SD) can cause a decline in learning and memory and lead to changes in behavior. Therefore, REM sleep plays a key role in processes that govern learning and memory. However, the mechanism underlying REM-SD-induced
Nikolaus A Watson et al.
Nature communications, 11(1), 1684-1684 (2020-04-05)
There are thousands of known cellular phosphorylation sites, but the paucity of ways to identify kinases for particular phosphorylation events remains a major roadblock for understanding kinase signaling. To address this, we here develop a generally applicable method that exploits

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service