Skip to Content
Merck
All Photos(5)

Key Documents

ABN421

Sigma-Aldrich

Anti-Senataxin (OY11) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Probable helicase senataxin, Senataxin (OY11), Amyotrophic lateral sclerosis 4 protein, SEN1 homolog, Senataxin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$563.00

$563.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$563.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$563.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SETX(23064)

General description

Probable helicase senataxin (UniProt Q7Z333; also known as Amyotrophic lateral sclerosis 4 protein, SEN1 homolog) is encoded by the SETX (also known as ALS4, AOA2, SCAR1, bA479K20.2) gene (Gene ID 23064) in human. Senataxin is implicated in both genome and RNA transcriptome homeostasis. Senataxin contains a helicase domain for mediating RNA interaction and an N-terminal domain for critical protein interactions. Senataxin is reported to colocalize with DNA repair proteins, including the DNA damage response protein 53BP1, at distinct nuclear foci in S/G2-phase human cells, and the number of these foci increases in response to impaired DNA replication or DNA damage. Senataxin is shown to resolve RNA/DNA hybrids (R loops) that arise during transcription, which can lead to genome instability unless they are resolved efficiently.

Immunogen

A mixture of four KLH-conjugated linear peptides corresponding to sequences from both N- and C-terminal halves of human senataxin.

Application

Immunoprecipitation Analysis: 2 µg from a representative lot immunoprecipitated Senataxin from HeLa, U2OS, and HEK293 cell lysates (Courtesy of Dr. Kanagaraj Radhakrishnan and Dr. Stephen C. West, The Francis Crick Institute, London, UK).
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Senataxin and siRNA-mediated Senataxin downregulation in HEK293 cells (Courtesy of Dr. Kanagaraj Radhakrishnan and Dr. Stephen C. West, The Francis Crick Institute, London, UK).
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Flag-tagged Senataxin in anti-Flag immunoprecipitates from transfected HEK293 cells expressing Flag-tagged Senataxin (Courtesy of Dr. Kanagaraj Radhakrishnan and Dr. Stephen C. West, The Francis Crick Institute, London, UK).
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Senataxin in HEK293 cell lysates (Courtesy of Dr. Kanagaraj Radhakrishnan and Dr. Stephen C. West, The Francis Crick Institute, London, UK).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-Senataxin (OY11) Antibody is validated for use in Western Blotting and Immunoprecipitation for the detection of Senataxin .

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Senataxin in 10 µg of HUVEC lysate.

Target description

~300 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service