Skip to Content
Merck
All Photos(2)

Key Documents

ABN34

Sigma-Aldrich

Anti-PDE11A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Dual 3′,5′-cyclic-AMP and -GMP phosphodiesterase 11A, cAMP and cGMP phosphodiesterase 11A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$380.00

$380.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$380.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$380.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDE11A(50940)

General description

The PDE11A gene encodes for a dual-specificity Phosphodiesterase (PDE), which is expressed in the adrenal cortex and is partially inhibited by tadalafil and other PDE inhibitors. PDEs are known to catalyze cAMP and cGMP to 5’-AMP and 5’GMP, respectively and thereby regulate the intracellular concentration of these nucleotides. Loss-of-function mutation of the PDE11A gene is shown to increase cAMP signaling and cAMP-responsive element binding protein (CREB) phosphorylation, two conditions which are associated with adrenocortical hyperplasia, adenomas, and Cushing syndrome.

Specificity

This antibody recognizes GAF domain of PDE11A.

Immunogen

Epitope: GAF domain.
KLH-conjugated linear peptide corresponding to the GAF domain of human PDE11A.

Application

Detect PDE11A using this Anti-PDE11A Antibody validated for use in WB, IH.
Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected PDE11A in normal mouse midbrain tissue.
Research Category
Neuroscience
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in human hippocampus tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PDE11A in 10 µg of human hippocampus tissue lysate.

Target description

~104 kDa observed. Uniprot states four isoforms at a molecular weight of ~105 kDa, ~78 kDa, ~66 kDa, and ~56 kDa This antibody can only recognize the ~105 kDa and the ~78 kDa isoforms.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human hippocampus tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuromodulation targets intrinsic cardiac neurons to attenuate neuronally mediated atrial arrhythmias.
Gibbons, DD; Southerland, EM; Hoover, DB; Beaumont, E; Armour, JA; Ardell, JL
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service