Skip to Content
Merck
All Photos(2)

Key Documents

ABN2300

Sigma-Aldrich

Neuro-Chrom Pan Neuronal Marker Antibody-Rabbit

Neuro-Chrom, from rabbit

Synonym(s):

Neuronal Marker Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$650.00

$650.00


Check Cart for Availability


Select a Size

Change View
100 μL
$650.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$650.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

purified antibody

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Neuro-Chrom

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibodies to neuronal proteins have become critical tools for identifying neurons and discerning morphological characteristics in culture and complex tissue. While the labeling from classic histological techniques such as Golgi staining and modern molecular approaches such as GFP constructs yield excellent cytoarchitectural detail, these approaches are technically challenging and impractical for many neuroscience research needs. Neuron-specific antibodies are convenient precision tools useful in revealing cytoarchitecture, but are limited to the protein target distribution within the neuron, which may differ greatly from nucleus to soma to dendrite and axon. To achieve as complete a morphological staining as possible across all parts of neurons, Millipore has developed a polyclonal antibody blend that reacts against key somatic, nuclear, dendritic, and axonal proteins distributed across the pan-neuronal architecture that can then be detected by a single secondary antibody. This antibody cocktail has been validated in diverse methods, cell culture and immuno-histochemistry, giving researchers a convenient and specific qualitative and quantitative tool for studying neuronal morphology.

Specificity

Cat. # ABN2300 is specific to axons (neurites), dendrites, nucleus, and the cell body of neurons.
Reactivity with other species has not been determined.

Immunogen

Epitope: Whole Neuron Marker

Application

Neuro-Chrom Pan Neuronal Marker-Rabbit is an antibody targeting the Pan Neuronal Marker protein, validated for use in ICC, IHC & IF.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

Routinely tested Rat E18 cortex primary neurons in Immunocytochemistry.

Physical form

Blended polyclonal antibody cocktail in PBS with 0.05% NaN3
Format: Purified

Storage and Stability

Maintain at 2-8°C for up to 1 year from date of receipt.

Analysis Note

Control
Rat E18 cortex primary neuron cell culture or mouse adult brain cryosections.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey J Lochhead et al.
Scientific reports, 9(1), 2621-2621 (2019-02-24)
In the brain, insulin acts as a growth factor, regulates energy homeostasis, and is involved in learning and memory acquisition. Many central nervous system (CNS) diseases are characterized by deficits in insulin signaling. Pre-clinical studies have shown that intranasal insulin
Marina Mirchandani-Duque et al.
Biomedicines, 10(6) (2022-06-25)
Dysregulation of hippocampal neurogenesis is linked to several neurodegenereative diseases, where boosting hippocampal neurogenesis in these patients emerges as a potential therapeutic approach. Accumulating evidence for a neuropeptide Y (NPY) and galanin (GAL) interaction was shown in various limbic system
Cesar A Medina et al.
PloS one, 17(5), e0265604-e0265604 (2022-05-05)
Deterioration in the quality of a person's voice and speech is an early marker of Parkinson's disease (PD). In humans, the neural circuit that supports vocal motor control consists of a cortico-basal ganglia-thalamo-cortico loop. The basal ganglia regions, striatum and
Bas M J Olthof et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(5), 876-887 (2018-12-12)
Nitric oxide (NO) is a neurotransmitter synthesized in the brain by neuronal nitric oxide synthase (nNOS). Using immunohistochemistry and confocal imaging in the inferior colliculus (IC, auditory midbrain) of the guinea pig (Cavia porcellus, male and female), we show that
Marina A Silveira et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(24), 4685-4699 (2020-05-08)
Located in the midbrain, the inferior colliculus (IC) integrates information from numerous auditory nuclei and is an important hub for sound processing. Despite its importance, little is known about the molecular identity and functional roles of defined neuron types in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service