Skip to Content
Merck
All Photos(1)

Key Documents

ABN2251

Sigma-Aldrich

Anti-phospho NMDAR2A (Ser1459)

Synonym(s):

GluN2A, Glutamate [NMDA] receptor subunit epsilon-1, Glutamate receptor ionotropic NMDA 2A, N-methyl D-aspartate receptor subtype 2A, NR2A, hNR2A

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$233.00
100 μG
$590.00

$233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$233.00
100 μG
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$233.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

species reactivity

human

packaging

antibody small pack of 100

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

Gene Information

human ... GRIN2A(2903)

Specificity

This rabbit polyclonal antibody specifically detects Phospho-NMDAR2A (Ser1459).

Immunogen

A peptide corresponding to 176 amino acids, phosphorylated on Serine 1459, from the C-terminal region of Human NMDAR2A.

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from wild-type HEK293T cells.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NMDAR2A phosphorylated on serine 1459 in lysate from HEK293T cells, but not in lysates from cells transfected with S1459A NMDAR2A.

Tested Applications

Western Blotting Analysis: A representative lot detected phospho NMDAR2A (Ser1459) in Western Blotting applications (Wyklicky, H., et. al. (1988). Wien Klin Wochenschr. 100(7):208-10).

Immunocytochemistry Analysis: A representative lot detected phospho NMDAR2A (Ser1459) in Immunocytochemistry applications (Wyklicky, H., et. al. (1988). Wien Klin Wochenschr. 100(7):208-10).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Glutamate receptor ionotropic, NMDA2A (UniProt: Q12879; also known as GluN2A, Glutamate [NMDA] receptor subunit epsilon-1, N-methyl D-aspartate receptor subtype 2A, NMDAR2A, NR2A, hNR2A) is encoded by the GRIN2A (also known as NMDAR2A) gene (Gene ID: 2903) in human. NMDA receptor complexes are ligand-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. They are involved in long-term potentiation and neuronal plasticity and regulate the number of nerve cells during development. However, over-stimulation NMDA receptors can lead to neuronal degeneration. NMDAR2A is a multi-pass membrane glycoprotein that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off. NMDAR2A exists as a heterotetramer and the heterotetrameric channels are composed of two zeta subunits (GRIN1) and two epsilon subunits (GRIN2A, GRIN2B, GRIN2C or GRIN2D) (in vitro). It can also form heterotetrameric channels that contain at least one zeta subunit (GRIN1), at least one epsilon subunit, plus GRIN3A or GRIN3B. In vivo, the subunit composition may depend on the expression levels of the different subunits. These heterotetrameric, ligand-gated ion channels display high calcium permeability and voltage-dependent sensitivity to magnesium. Channel activation requires binding of the neurotransmitter glutamate to the epsilon subunit, glycine binding to the zeta subunit, plus membrane depolarization to eliminate channel inhibition by magnesium ions. It has been shown that channels containing GRIN1 and GRIN2A have higher sensitivity to glutamate and faster kinetics than channels formed by GRIN1 and GRIN2B. Mutations in GRIN2A gene are known to cause epilepsy, speech disorders, and mental retardation. Two isoforms of NMDAR2A have ben described that are produced by alternative splicing. (Ref.: Hess, SD. et al. (1996). J. Pharmacol. Exp. Therap. 278(2); 808-816).

Physical form

Purified rabbit polyclonal antibody in buffer containing PBS, pH 7.4, containing no preservatives.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service