Skip to Content
Merck
All Photos(2)

Documents

ABN20

Sigma-Aldrich

Anti-Dopamine D(1A) Receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

dopamine receptor D1, D(1A) dopamine receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Drd1(24316)

General description

Dopaminergic signals are thought to modulate corticostriatal synapses onto medium spiny projection neurons, and this plasticity is, in turn, thought to alter patterns of activity during learning. Subversion of these processes in clinical and rodent models is hypothesized to contribute importantly to addiction to psychostimulants, seizures, and movement disorders in Huntington’s (HD) and Parkinson’s disease (PD). Dopamine receptor D1, also known as DRD1, is the most abundant dopamine receptor (of the five types D1-D5) in the central nervous system and is critically involved in these processes and the modulation of D2 receptor mediated events. Coupled with G-protein, the D1a receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases including the key molecular switch, DARPP-32.

Specificity

This antibody recognizes D(1A) dopamine receptor.

Immunogen

Epitope: Cytoplasmic Domain
KLH-conjugated linear peptide of rat dopamine D(1A) receptor at and around the cytoplasmic domain.

Application

Anti-Dopamine D(1A) Receptor Antibody detects level of Dopamine D(1A) Receptor & has been published & validated for use in IH, WB, ELISA.
Immunohistochemistry Analysis: A 1:300 dilution from a previous lot detected D(1A) dopamine receptor in rat brain tissue.

ELISA Analysis: Tested in ELISA against antigen-coated plate. A 1:100 dilution from a previous lot detected dopamine D(1A) receptor peptide.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Evaluated by Western Blot in rat testis tissue lysate.

Western Blot Analysis: 2 µg/ml of this antibody detected dopamine D(1A) receptor in 10 µg of rat testis tissue lysate.

Target description

~ 55 kDa is observed

Linkage

Replaces: AB1765P;

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat testis tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory N Ruegsegger et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(12), 5371-5383 (2017-08-11)
Prenatal overnutrition affects development into adulthood and influences risk of obesity. We assessed the transgenerational effect of maternal Western diet (WD) consumption on offspring physical activity. Voluntary wheel running was increased in juvenile (4-7 wk of age), but decreased in
Silvana Obici et al.
Molecular metabolism, 4(10), 692-705 (2015-10-27)
Melanocortin-4 receptors (MC4Rs) are highly expressed by dopamine-secreting neurons of the mesolimbic tract, but their functional role has not been fully resolved. Voluntary wheel running (VWR) induces adaptations in the mesolimbic dopamine system and has a myriad of long-term beneficial
Cell-type specific increases in female hamster nucleus accumbens spine density following female sexual experience.
Staffend, NA; Hedges, VL; Chemel, BR; Watts, VJ; Meisel, RL
Brain Structure &Amp; Function null
Dafna Lotan et al.
PloS one, 9(6), e101257-e101257 (2014-07-01)
Post-streptococcal A (GAS) sequelae including movement and neuropsychiatric disorders have been associated with improvement in response to antibiotic therapy. Besides eradication of infection, the underlying basis of attenuation of neuropsychiatric symptoms following antibiotic treatment is not known. The aim of
Andrea Stojakovic et al.
Brain sciences, 11(4) (2021-04-04)
Evidence suggests that nicotine and alcohol can each serve as a gateway drug. We determined whether prior nicotine and alcohol treatment would alter amphetamine reward. Also, we examined whether age and dopaminergic neurotransmission are important in this regard. Male and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service