Skip to Content
Merck
All Photos(2)

Key Documents

ABN1687

Sigma-Aldrich

Anti-TBR-2

from rabbit

Synonym(s):

Eomesodermin homolog, T-box brain protein 2, T-brain-2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00
100 μG
$545.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00
100 μG
$545.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Eomes(13813)

General description

Eomesodermin homolog (UniProt: O54839; also known as T-box brain protein 2, T-brain-2, TBR-2) is encoded by the Eomes (also known as Tbr2) gene (Gene ID: 13813) in murine species. T-box genes are a family of transcription factors have a highly conserved DNA binding domain named the T-domain. TBR-2 functions as a transcriptional activator that plays a crucial role during gastrulation, mesodermal specification, and limb patterning. It plays a role in brain development being required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex. It is also shown to be involved in the differentiation of CD8+ T-cells during immune response regulating the expression of lytic effector genes. Its DNA-binding region is localized to amino acids 278-458. Conditional inactivation of TBR-2 during early brain development is known to cause microcephaly and severe behavior deficits. Two isoforms of TBR-2 have been described that are produced by alternative splicing. Isoform 2 lacks amino acids 463-481. (Ref.: Bulfone, A., et al. (1999). Mech. Dev. 84 (1-2); 133-138).

Specificity

This rabbit polyclonal antibody detects T-box brain protein 2 (TBR-2) in murine species. It targets an epitope with in 20 amino acids from the C-terminal region.

Immunogen

Conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine TBR-2.
Epitope: C-terminus

Application

Anti-TBR-2, Cat. No. ABN1687, is a highly specific rabbit polyclonal antibody that targets Eomesodermin homolog (Tbr2) and has been tested in Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected TBR-2 in Immunofluorescence applications (Nelson, B.R., et. al. (2013). J Neurosci. 33(21):9122-39; Hodge, R.D., et. al. (2013). J Neurosci. 33(9):4165-80).

Immunohistochemistry Analysis: A representative lot detected TBR-2 in Immunohistochemistry applications (Yoon, K.J., et. al. (2008). Neuron. 58(4):519-31).

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TBR-2 in mouse hippocampus, embryonic mouse brain, and embryonic mouse gut tissue sections.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in mouse E13-14 brain.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TBR-2 in mouse E13-14 brain.

Target description

~63 kDa observed; 74.80 kDa calculated.

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura González-Blanco et al.
Cells, 10(10) (2021-10-24)
The 18-kDa translocator protein (TSPO) is a key mitochondrial target by which different TSPO ligands exert neuroprotective effects. We assayed the neurogenic potential of TSPO to induce the neuronal differentiation of pluripotent P19 stem cells in vitro. We studied changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service