Skip to Content
Merck
All Photos(1)

Key Documents

ABN1672

Sigma-Aldrich

Anti-HD-polyAla Ct

serum, from rabbit

Synonym(s):

HD-RAN-PolyAla, HD-PolyAla

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$277.00
100 μL
$700.00

$277.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$277.00
100 μL
$700.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$277.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

target post-translational modification

unmodified

General description

Huntington disease (HD) is a progressive neurodegenerative disorder is caused by a CAG,CTG expansion in the HTT gene that results in the production of a mutant huntingtin protein (HTT) with polymeric expansions that accumulate in human brain with HD. PolyAla is a sense repeat-associated non-ATG (RAN) translation protein that accumulates in various brain regions with some toxic effects leading to microglial activation and neuronal loss. HD-RAN polyAla toxicity is shown to be lower compared to that of polyGln. The transfection of cells with polyAla construct can express high polyAla levels, but have lower toxicity. HD-polyAla RAN proteins are reported to accumulate in cells expressing CAG repeats that are equal to or greater than 52 but not at repeats less than 45. HD-RAN proteins are abundant in regions of the brain that are most affected by HD and these regions display pathologic features of HD, such as caspase-3 activity and microglial activation. HD-Ran protein accumulation and aggregation in HD brains are shown to be length dependent.

Specificity

This rabbit polyclonal antibody specifically detects RAN proteins with PolyAla.

Immunogen

A synthetic peptides from the C-terminal regions of the predicted 21 polyAla frame of HD in the CAG direction.

Application

Anti-HD-polyAla Ct, Cat. No. ABN1672, is a highly specific rabbit polyclonal antibody that targets HD-polyAla Ct and has been tested in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected HD-polyAla Ct in Immunohistochemistry applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Western Blotting Analysis: A representative lot detected HD-polyAla Ct in Western Blotting applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Immunocytochemistry Analysis: A representative lot detected HD-polyAla Ct in Immunocytochemistry applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HD-polyAla Ct in pcDNA3.1 (Ctrl) & HEK293T-HD-polyAla-Ct.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293T cells transfected with HD-polyAla-Ct.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected PolyAla in HEK293T cells transfected with HD-polyAla-Ct.

Target description

~15 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service