Skip to Content
Merck
All Photos(3)

Documents

ABN1669M

Sigma-Aldrich

Anti-Anoctamin-1 (TMEM16A)

from rabbit

Synonym(s):

Anoctamin-1, TMEM16A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ANO1(55107)
mouse ... Ano1(101772)

General description

Anoctamin-1 (UniProt: Q8BHY3; also known as Transmembrane protein 16A, TMEM16A) is encoded by the Ano1 (also known as Tmem16a) gene (Gene ID: 101772) in murine species. Anoctamin-1 is a multi-pass membrane protein that along with TMEMB16B subunit serves as the calcium-activated chloride channels (CaCC). Two isoforms of Anoctamin-1 have been reported that are produced by alternative splicing. It is located in the apical membranes of epithelial cells in exocrine glands and in trachea. Its airway epithelial expression is increased in asthmatics. Hence, its abundance and activity may contribute to mucus secretion and airway hyper-responsiveness in asthma subjects. Anoctamin-1 is reported to be absent from smooth muscle cells, but present in the interstitial cells of Cajal (ICC), the pacemaker cells that control smooth muscle contraction. It acts as a major contributor to basal and stimulated chloride conductance in airway epithelial cells and plays an important role in tracheal cartilage development. ATP and calmodulin are shown to be essential for its activation and the channel activity is inhibited by CFTR protein and by chloride channel inhibitors such as niflumic acid and 4, 4 -diisothiocyanatostilbene-2,2′-disulfonic acid (DIDS). Mice with defective Tmem16a gene display severe tracheomalacia with gaps in the tracheal cartilage rings along the entire length of the trachea and over 90% of mutants die within the first nine days of postnatal life.

Specificity

This rabbit polyclonal antibody detects Anoctamin-1/TMEM16A in human and murine cells. it targets an epitope within 14 amino acids from the N-terminal end. May react with both isoforms 1 and 2 based on sequence homology.

Immunogen

A linear peptide corresponding to 14 amino acids from the N-terminal region of murine Anoctamin-1 (TMEM16A).
Epitope: N-terminus

Application

Anti-Anoctamin-1 (TMEM16A), Cat. No. ABN1669, is a highly specific rabbit polyclonal antibody that targets Anoctamin-1 (Transmembrane protein 16A) and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting..
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Anoctamin-1 (TMEM16A) in 10 µg of SW-620 and PC3 cell lysates.

Immunofluorescence Analysis: A representative lot detected Anoctamin-1 (TMEM16A) in Immunofluorescence applications (Huang, F., et. al. (2009). Proc Natl Acad Sci USA. 106(50):21413-8; Wang, H.C., et. al. (2015). Cell. 163(6):1348-59; Huang, F., et. al. (2012). Proc Natl Acad Sci USA. 109(40):16354-9).

Immunocytochemistry Analysis: A representative lot detected Anoctamin-1 (TMEM16A) in Immunocytochemistry applications (Huang, F., et. al. (2009). Proc Natl Acad Sci USA. 106(50):21413-8).

Western Blotting Analysis: A representative lot detected Anoctamin-1 (TMEM16A) in Western Blotting applications (Huang, F., et. al. (2009). Proc Natl Acad Sci USA. 106(50):21413-8).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Anoctamin-1 (TMEM16A) in 10 µg of A431 cell lysate.

Target description

~111 kDa observed; 110.92 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service