Skip to Content
Merck
All Photos(2)

Documents

ABN1477

Sigma-Aldrich

Anti-nSR100 (SRRM4)

from rabbit

Synonym(s):

Serine/arginine repetitive matrix protein 4, Medulloblastoma antigen MU-MB-2.76, Neural-specific serine/arginine repetitive splicing factor of 100 kDa, Neural-specific SR-related protein of 100 kDa, nSR100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SRRM4(84530)

General description

Serine/arginine repetitive matrix protein 4 (UniProt: A7MD48; also known as Medulloblastoma antigen MU-MB-2.76, Neural-specific serine/arginine repetitive splicing factor of 100 kDa, Neural-specific SR-related protein of 100 kDa, nSR100) is encoded by the SRRM4 (also known as KIAA1853) gene (Gene ID: 84530) in human. nSR100 is a nuclear protein that is specifically expressed in neuronal cells. It serves as a splicing factor that is specifically required for neural cell differentiation. It acts in conjunction with nPTB/PTBP2 by binding directly to its regulated target transcripts and promote neural-specific exon inclusion in many genes that function in neural cell differentiation. It is shown to regulate the splicing of protrudin gene (Zfyve27) transcripts in neuronal cells and promotes splicing of protrudin pre-mRNA so as to include a microexon (exon L) encoding seven amino acids in a neuron-specific manner. The resulting protein known as Protrudin-L promotes neurite outgrowth during neurogenesis. Depletion of nSR100 in Neuro2A cells is shown to impair neurite outgrowth, whereas its ablation in mouse embryos is reported to prevent differentiation of neuronal progenitors in the cortex and impair development of the central and peripheral nervous systems. (Ref.: Ohnishi, T et al. (2017). Sci. Rep. 7: 41130; Quesnel-Vallières, M et al. (2015). Genes Dev. 29(7); 746-759).

Specificity

This rabbit polyclonal antibody detects Serine/arginine repetitive matrix protein 4 in human and mouse cells. It targets an epitope within 104 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant frgment corresponding to 104 amino acids from the C-terminal half of human Serine/arginine repetitive matrix protein 4.

Application

Anti-nSR100 (SRRM4), Cat. No. ABN1477, is a highly specific rabbit polyclonal antibody that targets Serine/arginine repetitive matrix protein 4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected nSR100 (SRRM4) in mouse cerebral cortex and human cerebral cortex tissue sections.

Quality

Evaluated by Western Blotting in human cerebellum tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected nSR100 (SRRM4) in 10 µg of human cerebellum tissue lysate.

Target description

~68 kDa observed; 68.56 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service