Skip to Content
Merck
All Photos(2)

Key Documents

ABN1346

Sigma-Aldrich

Anti-SHIP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 1, hp51CN, Inositol polyphosphate-5-phosphatase of 145 kDa, p150Ship, SH2 domain-containing inositol 5′-phosphatase 1, SH2 domain-containing inositol phosphatase 1, SHIP-1, SHIP1, SIP-145

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$449.00

$449.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$449.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$449.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... INPP5D(3635)

General description

Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 1 (EC 3.1.3.86; UniProt Q92835; also known as hp51CN, Inositol polyphosphate-5-phosphatase of 145 kDa, p150Ship, SH2 domain-containing inositol 5′-phosphatase 1, SH2 domain-containing inositol phosphatase 1, SHIP-1, SIP-145) is encoded by the INPP5D (also known as SHIP, SHIP1) gene (Gene ID 3635) in human. SHIP1 and SHIP2 are related phosphatases that play important roles in modulating cell signaling by regulating the phosphorylation state of phosphatidylinositol phosphates (PIPs). While SHIP1 and SHIP2 share a high degree of amino acid conservation, they differ significantly in their cellular expression and receptor recruitment. SHIP1 is primarily confined to cells of the hematopoietic lineage, although it is also expressed by embryonic and mesenchymal stem cells (ESCs and MSCs). SHIP2 is ubiquitously expressed in all cell and tissue types, with especially high levels found in the heart, skeletal muscle, and placenta. SHIP1 functions as s negative regulator in immunoreceptor signaling, in hematopoietic progenitor cell proliferation/survival, and as an inducer of cellular apoptosis. Humna SHIP1 consists of an N-terminal SH2 domain (a.a. 5-101) that mediates interactions with a large number of intracellular signaling proteins. SHIP1 also contains three SH3-interacting regions (a.a. 124-129, 969-974, 1040-1051) characterized by the presence of the PxxP motifs and two NPXY motifs (a.a. 912-915 & 1019-1022) that interact with SH2 domain-containing proteins.

Specificity

Immunogen sequence is present in all three spliced isoforms of human SHIP1 reported by UniProt (Q92835).

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to a sequence near the C-terminus of human SHIP1.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected SHIP1 in human lymphoma tissue, as well as in normal tonsil and kidney tissue sections.
Research Category
Neuroscience
Research Sub Category
Kinases & Phosphatases
This Anti-SHIP1 Antibody is validated for use in Western Blotting, Immunohistochemistry for the detection of SHIP1.

Quality

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected SHIP1 in 10 µg of Ramos cell lysate.

Target description

~145 kDa observed. Target band size appears larger than the calculated molecular weights of 133.63 kDa (isoform 1), 133.2 kDa (isoform 2), 109.3 kDa (isoform 3) due to posttranslational phosphorylation.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service