Skip to Content
Merck
All Photos(3)

Key Documents

ABF131

Sigma-Aldrich

Anti-phospho TIE2/TEK Antibody (Tyr992)

0.25 mg/mL, from rabbit

Synonym(s):

Angiopoietin-1 receptor, Endothelial tyrosine kinase, Tunica interna endothelial cell kinase, Tyrosine kinase with Ig and EGF homology domains-2, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, hTIE2, p140 TEK, CD202b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$641.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), zebrafish (based on 100% sequence homology), bovine (based on 100% sequence homology)

concentration

0.25 mg/mL

technique(s)

immunoprecipitation (IP): suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr992)

Gene Information

human ... TEK(7010)

General description

TIE2/TEK (tyrosine kinase with Ig and EGF homology domains 2) is expressed almost exclusively in endothelial cells in mice, rats and humans. This receptor possesses a unique extracellular domain containing two immunoglobulin-like loops separated by three epidermal growth factor-like repeats that are connected to three fibronectin type III-like repeats. The ligand for the receptor is Angiopoietin 1. Defects in TIE2/TEK are associated with inherited venous malformations; the TIE2/TEK signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis.

Immunogen

Epitope: Protein Kinase Domain
KLH-conjugated linear peptide corresponding to the Protein Kinase Domain of human TIE2/TEK phosphorylated at Tyr992.

Application

Immunoprecipitation Analysis: 5 µL from a representative lot immunoprecipitated phospho TIE2/TEK (Tyr992) from Pervanadate treated HUVEC cell lysate. Immunoprecipated sample was then subjected to Western Blotting using a 1:5,000 dilution of this antibody.

Peptide Inhibition Assay Analysis: A 1:500 dilution from a representative lot detected phospho TIE2/TEK (Tyr992) from transfected HEK293T lysates when pre-incubated without or with non-phospho peptide, and not when pre-incubated with phospho peptide.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
This Anti-phospho TIE2/TEK Antibody (Tyr992) is validated for use in western blotting, IP & peptide inhibition assay for the detection of phospho TIE2/TEK (Tyr992).

Quality

Evaluated by Western Blotting in untreated and Pervanadate treated HUVEC cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected phospho TIE2/TEK (Tyr992) in 10 µg of Pervanadate treated HUVEC cell lysate, and not in untreated HUVEC cell lysate.

Target description

~125 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Linkage

Replaces: 09-407

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah J Higgins et al.
The Journal of clinical investigation, 128(4), 1471-1484 (2018-01-24)
Disordered coagulation contributes to death in sepsis and lacks effective treatments. Existing markers of disseminated intravascular coagulation (DIC) reflect its sequelae rather than its causes, delaying diagnosis and treatment. Here we show that disruption of the endothelial Tie2 axis is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service