Skip to Content
Merck
All Photos(2)

Documents

ABF114

Sigma-Aldrich

Anti-MSR1/CD204 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Macrophage scavenger receptor types I and II, Macrophage acetylated LDL receptor I and II, Scavenger receptor class A member 1, CD204

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MSR1(4481)

Related Categories

General description

Macrophage scavenger receptors (MSR) are membrane glycoproteins that mediate the recognition and uptake of a wide variety of negatively charged macromolecules. There are two types of these receptors that differ in the C-terminal region. MSR has a key role in scavenging a variety of macromolecules, including modified lipoproteins, advanced glycation end (AGEs) products and amyloid b-protein (Abeta). While the normal role of MSR is associated with cell adhesion and host defense mechanisms, it also has been implicated in the development of atherosclerosis and Ab deposition in Alzheimer′s disease (AD).The observation that the 4 allele of the apolipoprotein E gene is associated with an increased risk for AD has lead to the investigation of lipoprotein metabolism in the brain of AD patients. The type I MSR has been found to be expressed on microglia and Mato cells, but not astrocytes or neurons. In AD brain, MSR type I is strongly expressed by microglia cells that are associated with senile plaques. Because the senile plaques contain AGEs, which are a source of free radicals that could result in oxidative damage to the brain, MSR may have a role in mediating the endocytosis of AGEs.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human MSR1/CD204.

Application

Anti-MSR1/CD204 is an antibody against MSR1/CD204 for use in WB, FC.
Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected MSR1/CD204 in 1X10E6 K562 myelogenous leukemia cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected MSR1/CD204 in 10 µg of THP-1 cell lysate.

Target description

~42 & ~49 kDa observed. This antibody is predicted to cross-react with all isoforms of MSR1/CD204 (Isoform 1 - ~49 kDa, Isoform 2 - ~40 kDa, and Isoform 3 - ~42 kDa).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service