Skip to Content
Merck
All Photos(1)

Key Documents

ABF1066

Sigma-Aldrich

Anti-SARS-CoV-2 spike protein ectodomain (S1+S2)

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$300.00
100 μL
$753.00

$300.00


Estimated to ship on01 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZMS1076

Request a Bulk Order

Select a Size

Change View
25 μL
$300.00
100 μL
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$300.00


Estimated to ship on01 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZMS1076

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody
serum

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 139.13 kDa
observed mol wt ~170 kDa

purified by

(Unpurified)

species reactivity

virus

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
neutralization: suitable
western blot: suitable

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

vaccinia virus ... s> S(43740568)

General description

Spike glycoprotein (UniProt: P0DTC2; also known as S glycoprotein, E2, Peplomer protein) is encoded by the S (also known as 2) gene (Gene ID: 43740568) in Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). The SARS-CoV-2 is a positive-strand RNA virus that causes severe respiratory syndrome in human. The mature SARS-CoV-2 contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). E and M proteins help in viral assembly and N protein is needed for RNA synthesis. The S protein is a single-pass type I, homotrimeric, membrane glycoprotein that is responsible for virus binding and entry into host cell. It is synthesized with a signal peptide (aa 1-12), which is subsequently cleaved off to generate the mature protein that contains an extracellular domain (aa 13-1213), a transmembrane domain (aa 1214-1234), and a cytoplasmic domain 1235-1273). The S protein is further processed into S1 (aa 13-685) and S2 (aa 686-1273) subunits by host cell furin. The presence of a furin polybasic cleavage site in S protein from SARS-CoV-2 sets it apart from S protein in SARS-CoV that possesses a monobasic S1/S2 cleavage site. The S1 subunit has the receptor binding domain (RBD; aa 319-541) that mediates entry of SARS-CoV-2 into sensitive cells through the peptidase domain of host Angiotensin-converting enzyme 2 (ACE2) with high affinity (KD = 15 nM). The S2 domain (aa 686-1213), which is reported to be well conserved, is responsible for membrane fusion. This polyclonal antibody binds and neutralizes the SARS-CoV-2 variants including the B.1.1.7; B.1.429; P.1; and B.1.351 at different levels. (Ref.: Ravichandran, S., et al. (2020). Sci Transl Med. 12(550):eabc3539).

Specificity

This rabbit polyclonal antibody detects the extracellular domain (S1+ S2) of SARS-CoV-2 spike protein.

Immunogen

His-tagged recombinant fragment corresponding to the extracellular domain (S1 + S2; aa 16-1213) of SARS-CoV-2 spike protein.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant extracellular domain (S1+S2) of SARS-CoV-2 spike protein.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant extracellular domain (S1 + S2) of Spike glycoprotein of SARS-CoV-2.

Tested Applications

Enzyme Immunoassay (ELISA) Analysis: A representative lot detected SARS-CoV-2 spike protein ectodomain (S1+S2) in ELISA applications (Ravichandran, S., et. al. (2020). Sci Transl Med. 12(550):eabc3539).

Neutralizing: A representative lot neutralized SARS-CoV-2 spike protein ectodomain (S1+S2) in Neutralizing applications (Ravichandran, S., et. al. (2020). Sci Transl Med. 12(550):eabc3539).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SARS-CoV-2 spike protein ectodomain (S1+S2), Cat. No. ABF1066, is a rabbit polyclonal antibody that detects the extracellular domain (S1+ S2) of SARS-CoV-2 spike protein and is tested for use in ELISA, Neutralizing, and Western Blotting.

Physical form

Rabbit polyclonal antibody in serum without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service