Skip to Content
Merck
All Photos(1)

Key Documents

ABE601

Sigma-Aldrich

Anti-CPEB1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cytoplasmic polyadenylation element-binding protein 1, CPE-BP1, CPE-binding protein 1, h-CEBP, hCPEB-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$529.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CPEB1(64506)

General description

Cytoplasmic polyadenylation element-binding protein 1 (CPE-BP1, CPEB1) is an RNA binding protein that contains an RNA-recognition motif and a zinc finger-containing region and is found in a wide range of vertebrates and invertebrates. CPEB1 forms the nucleus of a complex of factors that regulate poly(A) elongation and promotes polyadenylation-induced translation. CPEB1 mediates many diverse biological processes such as germ cell development, cell division and senescence, and synaptic plasticity. Recently, it was discovered that CPEB1 is involved in beta-catenin mRNA translation and cell migration in astrocytes as well as regulating hypoxia-inducible factor (HIF)-1 expression, demonstrating the wide range of processes in which CPEB1 plays a role. At least four isoforms of CPEB1 are known to exist.

Specificity

This antibody recognizes the N-terminus of CPEB1.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human CPEB1.

Application

Detect Cytoplasmic polyadenylation element-binding protein 1 using this rabbit polyclonal antibody, Anti-CPEB1 Antibody validated for use in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected CPEB1 in 15 µg of rat brain tissue lysate.

Target description

~65 kDa observed. Uniprot describes 2 isoforms at ~54 kDa, one isoform at ~62 kDa, and one isoform at ~63 kDa Uncharacterized bands may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service