Skip to Content
Merck
All Photos(1)

Key Documents

ABE305-S

Sigma-Aldrich

Anti-trimethyl Histone H3 (Lys36) Antibody, Trial Size

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l, H3k36me3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μL
$219.00

$219.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 μL
$219.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$219.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

ChIP: suitable
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys36)

Gene Information

human ... HIST1H3F(8968)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the "histone code" hypothesis). The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Methylation of histone H3 on Lys36 (H3K36me2/3) is tightly associated with actively transcribed genes, and this modification is found primarily within the coding region, suggesting H3K36 methylation is necessary for efficient RNA polymerase II elongation and processivity.

Specificity

This antibody recognizes Histone H3 trimethylated at Lys36.

Immunogen

Epitope: Trimethylated Lys36
Linear peptide corresponding to human Histone H3 trimethylated at Lys36.

Application

Dot Blot Analysis: A 1:500 dilution from a representative lot detected trimethyl Histone H3 (Lys36).

Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in mouse embryonic stem cells. (Laboratory of Dr. Rob Klose, Oxford University)

Immunofluorescence Analysis: A representative lot was used by an independent laboratory in NIH/3T3 fibroblasts. (Laboratory of Dr. Rob Klose, Oxford University)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-trimethyl Histone H3 (Lys36) Antibody (rabbit polyclonal antibody), Trial Size/ Pack validated in WB, DB, ICC, ChIP to detect trimethyl Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).

Quality

Evaluated by Western Blot in Histone H3 recombinant proteins.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected Histone H3 on 10 µg of Histone H3 recombinant proteins.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Histone H3 recombinant proteins

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service