Skip to Content
Merck
All Photos(1)

Key Documents

ABE278

Sigma-Aldrich

Anti-MILI Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Piwi-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$578.00

$578.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$578.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$578.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

MILI, MIWI, and MIWI2 belong to the Piwi family of proteins. piRNAs and Piwi proteins are implicated in transposon control and are linked to transposon methylation in mammals. MILI, in particular, directly binds with piRNAs in the cytoplasm and is required for PIWIL4/MIWI2 nuclear localization and association with secondary piRNAs antisense. MILI plays an essential role in spermatogenesis, meiotic differentiation of spermatocytes, germ cell differentiation and in self-renewal of spermatogonial stem cells.

Immunogen

KLH-conjugated linear peptide corresponding to mouse MILI.

Application

Anti-MILI Antibody is a Rabbit Polyclonal Antibody for detection of MILI also known as Piwi-like protein 2 & has been validated in WB, ICC, IHC & IP.
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in male primordial germ cells. (Aravin, A.A., et al. (2008). Mol Cell. 31(6):785-799.)

Immunohistochemistry Fluoresence Analysis: A representative lot was used by an independent laboratory in embyronic testis cross sections. (Vagin, W., et al. (2009). Genes Dev. 23(15):1749-1762.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory. (Aravin, A.A., et al. (2007). Science. 316(5825):744-747.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in RAW264.7 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected MILI in 10 µg of RAW264.7 cell lysate.

Target description

~109 kDa observed. An uncharacterized bands at ~30, 55, and 60 kDa may be observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
RAW264.7 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service