Skip to Content
Merck
All Photos(6)

Key Documents

ABE219

Sigma-Aldrich

Anti-phospho Histone H3 (Ser57) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$683.00

$683.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer57)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

KLH-conjugated linear peptide corresponding to human Histone H3 phosphorylated at Ser57.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho Histone H3 (Ser57) in HeLa cells.

Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated phospho Histone H3 (Ser57) from 500 µg of colcemid treated HeLa acid extract.

Dot Blot (Specificity) Analysis: 0.005 µg/mL from a representative lot detected phospho Histone H3 (Ser57) in a panel containing various modified and non-modified Histone H3 peptides.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H3 (Ser57) Antibody is validated for use in western blotting, ICC, IP & dot blot for the detection of phospho Histone H3 (Ser57).

Quality

Evaluated by Western Blotting in colcemid treated HeLa acid extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected phospho Histone H3 (Ser57) in 10 µg of colcemid treated HeLa acid extract.

Target description

~17 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Sun et al.
EBioMedicine, 43, 171-179 (2019-05-08)
Diffuse intrinsic pontine glioma (DIPG) is a rare and fatal pediatric brain cancer without cure. Seeking therapeutic strategies is still a major challenge in DIPG research. Previous study has shown that dysregulation of G1/S cell cycle checkpoint was common in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service