Skip to Content
Merck
All Photos(1)

Key Documents

ABE1974

Sigma-Aldrich

Anti-CHMP1B

from rabbit, purified by affinity chromatography

Synonym(s):

Charged multivesicular body protein 1b, CHMP1.5, Chromatin-modifying protein 1b, Vacuolar protein sorting-associated protein 46-2, Vps46-2, hVps46-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$574.00

$574.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$574.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$574.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 98% homology), bovine (based on 99% homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CHMP1B(57132)

General description

Charged multivesicular body protein 1b (UniProt: Q7LBR1; CHMP1.5, Chromatin-modifying protein 1b, CHMP1b, Vacuolar protein sorting-associated protein 46-2, Vps46-2, hVps46-2) is encoded by the CHMP1B (also known as C18orf2) gene (Gene ID: 57132) in human. CHMP1b is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. It is widely expressed and is found in pancreas, kidney, skeletal muscle, liver, lung, placenta and the brain tissue. CHMP1b is a peripherally associated component of the endosomal sorting system that is required for transport complex III (ESCRT-III), which is involved in multi-vesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. CHMP1b is localized in cytosol, endosome, and in peripheral membrane protein. CHMP1b binds specifically to the AAA ATPase spastin and thereby recruits this microtubule severing protein to the midbody. CHMP1b and spastin are shown to colocalize at midbodies in dividing HeLa cells, and the knockdown of CHMP1b reduces the amount of spastin at midbodies.

Specificity

This polyclonal antibody detects Charged multivesicular body protein 1b in human cells. Human CHMP1B displays 99% and 98% homology with bovine and mouse CHMP1B, respectively.

Immunogen

Epitope: unknown
GST-tagged full length recombinant human Charged multivesicular body protein 1b.

Application

Anti-CHMP1B, Cat. No. ABE1974, is a highly specific rabbit polyclonal antibody that targets charged multivesicular body protein 1b and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected CHMP1B in Western Blotting applications (Morita, E., et. al. (2010). Proc Natl Acad. Sci USA. 107(29):12889-94; Bailey, J.K., et. al. (2015). J Biol Chem. 290(14):8987-9001).

Quality

Evaluated by Western Blotting in ARPE-19 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected CHMP1B in 10 µg of ARPE-19 cell lysate.

Target description

~29 kDa observed; 22.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service