Skip to Content
Merck
All Photos(1)

Key Documents

ABE1963

Sigma-Aldrich

Anti-RecQ5 Antibody

serum, from rabbit

Synonym(s):

ATP-dependent DNA helicase Q5, DNA helicase, RecQ-like type 5, RecQ5, RecQ protein-like 5

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$422.00

$422.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$422.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$422.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RECQL5(9400)

General description

ATP-dependent DNA helicase Q5 (EC 3.6.4.12; UniProt O94762; also known as DNA helicase RecQ-like type 5, RecQ5, RecQ protein-like 5) is encoded by the RECQL5 (also known as RECQ5) gene (Gene ID 9400) in human. Human RECQ proteins (RECQ1, BLM, WRN, RECQ4, and RECQ5) constitute a family of highly conserved DNA helicases with nonredundant functions to ensure proper development. RECQ5 plays an important role in preventing both transcription-associated double-stranded breaks (DSBs) and DSBs caused by TOP1 poisons, such as camptothecin (CPT), and Recq5 deficiency leads to genome instability and cancer development in mice. RECQ5 KIX domain mediated interaction with RNA polymerase II (RNAPII) prevents RNAPII from binding transcription elongation factor TFIIS, thereby suppressing RNAPII-dependent transcription initiation and DSBs. On the other hand, RECQ5 is also known to interact with the active RNAPII (RNAPIIo) elongation complex to maintain the transcription elongation rate and prevent genome instability. In addition, RECQ5 helicase domain mediated interaction with TOP1 is reported to facilitate TOP1 K391/K436 SUMOylation, promoting TOP1-RNAPIIo association and efficient recruitment of mRNA processing factors to the transcriptionally active chromatin to prevent genome instability caused by R-loops formation.

Specificity

This antiserum recognizes RECQ5 isoform Beta by targeting a sequence from the RAD51-interacting region within the helicase domain. This antiserum is expected to react with also spliced isoform 4, but not the helicase domain-truncated Alpha (RecQ5b) and Gamma (RecQ5a) spliced variants reported by UniProt (O94762).

Immunogen

Epitope: RAD51 interaction domain.
Linear peptide corresponding a sequence from the RAD51 interaction domain of human RecQ5.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-RecQ5 Antibody is validated for use in Western Blotting for the detection of RecQ5.
Western Blotting Analysis: A representative lot detected FLAG-tagged wild-type RecQ5 and a D157A (DA) ATPase-defective RecQ5 mutant construct, but not a helicase domain-truncated (a.a. 450–991 deleted) RECQ5 construct expressed in HEK293 T cells devoid of endogenous RecQ5 (Li, M., et al. (2015). Nat. Commun. 6:6720).
Western Blotting Analysis: Representative lots detected siRNA-mediated RecQ5 downregulation using whole cell lysate or chromatin fraction from human HEK293T cells. Combined treatment with siRNA and the transcription inhibitor 5,6-dichloro-beta-d-ribofuranosyl benzimidazole (DRB) further decreased cellular RecQ5 protein level (Li, M., et al. (2015). Nat. Commun. 6:6720; Li, M., et al. (2011). Mol. Cell. Biol. 31(10) 2090-2099).

Quality

Evaluated by Western Blotting in Raji nuclear extract.

Western Blotting Analysis: An 1:500 dilution of this antibody detected RecQ5 in 10 µg of Raji nuclear extract.

Target description

~120 kDa observed. 108.9 kDa (isoform Beta) and 106.1 kDa (isoform 4) calculated.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service